Order Entry
Northern Ireland
ContactUsLinkComponent
Triphenylphosphine selenide for synthesis, Sigma-Aldrich®
Triphenylphosphine selenide for synthesis, Sigma-Aldrich®
Catalog # 8.18054.0050
Supplier:  Merck
CAS Number:  
Triphenylphosphine selenide for synthesis, Sigma-Aldrich®
Catalog # 8.18054.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    186 - 189 °C
  • Molar Mass:
    341.25 g/mol
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

About this item

 :
 :341.25 g/mol
EINECS:223-406-1
UN:3467
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:3878-44-2
Formula:C₁₈H₁₅PSe
MDL Number:MFCD00013477
Melting Pt:186…189 °C
ADR:6.1 II
 :M-Clarity™ quality level = MQ200