Phosphoryl trichloride
MOLE32635781-1KG
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:153.33 g/mol
:12,07506
:107 °C (1013 hPa)
:1 °C
:1,645 g/cm³ (20 °C)
:Ambient
:233-046-7
:1810
:8,I
:GHS06
:GHS05
:GHS08
:10025-87-3
:POCl₃
:MFCD00011443