Phosphorus(III) chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7719-12-2
:
Formula:Cl₃P
MDL Number:MFCD00011438
:137.33 g/mol
Merck Index:12,07515
Boiling Pt:76 °C (1013 hPa)
Melting Pt:–112 °C
Density:1,57 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1809
ADR:6.1 (8),I
:M-Clarity™ quality level = MQ200
EINECS:231-749-3
:GHS06
:GHS05
:GHS08
Frequently Bought Together