Order Entry
Northern Ireland
ContactUsLinkComponent
Antimony(III) chloride ≥99.0%, AnalaR NORMAPUR® analytical reagent
Antimony(III) chloride ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 21470.186
Supplier:  VWR Chemicals
CAS Number:  
undefined
Antimony(III) chloride ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 21470.186
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Insolubility in chloroform:
    Max. 0.01 %
  • Insolubility in HCl:
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 20 ppm
  • K (Potassium):
    Max. 0.01 %
  • Na (Sodium):
    Max. 50 ppm
  • Pb (Lead):
    Max. 30 ppm

Specification Test Results

About this item

 :
 :228.12 g/mol
Merck Index:13,00713
Boiling Pt:223,5 °C (1013 hPa)
Melting Pt:73 °C
Density:3.14 g/cm³ (20 °C)
Density:3,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-047-2
UN:1733
ADR:8,II
 :GHS05
 :GHS07
 :GHS09
CAS Number:10025-91-9
Formula:SbCl₃
MDL Number:MFCD00011212