Antimony(III) chloride for synthesis, Sigma-Aldrich®
: Merck
:10025-91-9
:MFCD00134030
:SbCl₃
:MFCD00011212
:228.12 g/mol
:13,00713
:223,5 °C (1013 hPa)
:73 °C
:3,14 g/cm³ (20 °C)
:1733
:8,II
:M-Clarity™ quality level = MQ200
:233-047-2
:GHS05
:GHS09
:Ambient
- Solubility:931 g/l (20 °C) Hydrolysis
- Melting Point:73 °C
- Molar Mass:228.1 g/mol
- Bulk Density:1600 kg/m3
- Boiling Point:233 °C (1013 hPa)
- Vapor Pressure:0.16 hPa (20 °C)
- Density:3.14 g/cm3 (20 °C)
- pH:(H2O) acidic
- Synonyms:Antimonous acid, Antimony trichloride
- Hill Formula:Cl3Sb
- Chemical Formula:SbCl3
Frequently Bought Together