Phosphoryl tribromide
Catalog # MOLE76967164-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:286.69 g/mol
Merck Index:12,07505
Boiling Pt:193 °C (1013 hPa)
Melting Pt:52…55 °C
Density:2,88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-177-7
UN:1939
ADR:8,II
:GHS05
:GHS07
CAS Number:7789-59-5
Formula:Br₃OP
MDL Number:MFCD00036298