Order Entry
Northern Ireland
ContactUsLinkComponent
Phosphoryl tribromide for synthesis, Sigma-Aldrich®
Phosphoryl tribromide for synthesis, Sigma-Aldrich®
Catalog # 8.21026.1000
Supplier:  Merck
CAS Number:  
Phosphoryl tribromide for synthesis, Sigma-Aldrich®
Catalog # 8.21026.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    48 - 56 °C
  • Molar Mass:
    286.68 g/mol
  • Boiling Point:
    192 °C (1013 hPa) decomposes
  • Density:
    2.82 g/cm3
  • Synonyms:
    Phosphorus oxide tribromide, Phosphorus oxybromide
  • Hill Formula:
    Br3OP
  • Chemical Formula:
    POBr3

Specification Test Results

About this item

 :
 :286.69 g/mol
Merck Index:12,07505
Boiling Pt:193 °C (1013 hPa)
Melting Pt:52…55 °C
Density:2,88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-177-7
UN:1939
ADR:8,II
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS07
CAS Number:7789-59-5
Formula:Br₃OP
MDL Number:MFCD00036298