


Carbon disulphide 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-15-0
:
Formula:CS₂
MDL Number:MFCD00011321
:76.14 g/mol
Merck Index:13,01821
Boiling Pt:46 °C (760 mmHg)
Melting Pt:–111 °C
Density:1,262 g/cm³
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:200-843-6
UN:1131
ADR:3,
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 %
- GC:≥99.85 %
- UV at 380 nm:A: ≤1.3
- UV at 390 nm:A: ≤0.5
- UV at 400 nm:A: ≤0.1
- UV at 410 nm:A: ≤0.05
- UV at 420 nm:A: ≤0.01
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
Specification Test Results
Frequently Bought Together