

Tetrachlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10026-04-7
:
Formula:Cl₄Si
MDL Number:MFCD00011229
:169.9 g/mol
Merck Index:12,08644
Boiling Pt:57,6 °C (1013 hPa)
Melting Pt:–70 °C
Density:1,48 g/cm³ (20 °C)
UN:1818
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:233-054-0
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:-70 °C
- Molar Mass:169.90 g/mol
- Boiling Point:57 °C (1013 hPa)
- Vapor Pressure:260 hPa (20 °C)
- Density:1.48 g/cm3 (20 °C)
- pH:1 (5 g/l, H2O)
- Synonyms:Silicon(IV) chloride, Tetrachlorosilane
- Hill Formula:Cl4Si
- Chemical Formula:SiCl4
Specification Test Results
Related Information
Frequently Bought Together