Order Entry
Northern Ireland
ContactUsLinkComponent
Orthoboric acid ≥99.5%, crystallised, Gen-Apex for molecular biology
Orthoboric acid ≥99.5%, crystallised, Gen-Apex for molecular biology
Catalog # 33601.261
Supplier:  VWR Chemicals
CAS Number:  
undefined
Orthoboric acid ≥99.5%, crystallised, Gen-Apex for molecular biology
Catalog # 33601.261
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Colouration (0,1 mol/l; water):
    Max. 10 APHA
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Transmittance (230 nm) (0,1 mol/l):
    Min. 99 %
  • Transmittance (260 nm) (0,1 mol/l):
    Min. 95 %
  • Transmittance (280 nm) (0,1 mol/l):
    Min. 97 %
  • Transmittance (320 nm) (0.1 mol/l):
    Min. 99 %

Specification Test Results

About this item

 :
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1,44 g/cm³ (25 °C)
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
 :GHS08
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337