Orthoboric acid ≥99.5%, granular NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1,44 g/cm³ (25 °C)
:Management of Change (MOC) category = R
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
:GHS07
Specification Test Results
- Bulk Pharmaceutical Chemical:99.5 - 100.5 %
- NF - Assay (H₃BO₃):99.5 - 100.5 %
- NF - Completeness of Solution:Passes Test
- NF - Identification:Passes Test
- NF - Loss on Drying:≤0.5 %
- NF - Solubility in Alcohol:Passes Test
- Color of Solution:Passes Test
Specification Test Results
Frequently Bought Together