
Tetraethylene glycol dimethyl ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:143-24-8
:
Formula:C₁₀H₂₂O₅
MDL Number:MFCD00008505
:222.28 g/mol
:M-Clarity™ quality level = MQ200
EINECS:205-594-7
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-30 °C
- Molar Mass:222.28 g/mol
- Boiling Point:272 - 277 °C (1013 hPa)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:141 °C
- Refractive Index:1.4322 (20 °C, 589 nm)
- Explosion Limit:0.8 %(V)
- Density:1.01 g/cm3 (20 °C)
- Ignition Point:200 °C
- Viscosity kinematic:4.1 mm2/s (20 °C)
Specification Test Results
Frequently Bought Together