Order Entry
Northern Ireland
ContactUsLinkComponent
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Health
Irritant
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Catalog # 8.08249.0250
Supplier:  Merck
CAS Number:  
Health
Irritant
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Catalog # 8.08249.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -40 °C
  • Molar Mass:
    178.23 g/mol
  • Boiling Point:
    210 - 230 °C (1013 mbar)
  • Vapor Pressure:
    1.2 hPa (20 °C)
  • Flash Point:
    113 °C
  • Refractive Index:
    1.4233 (20 °C, 589 nm)
  • Explosion Limit:
    0.7 %(V)
  • Density:
    0.99 g/cm3 (20 °C)
  • pH:
    (H2O) neutral
  • Ignition Point:
    195 °C

Specification Test Results

About this item

 :
 :178.23 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:203-977-3
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:112-49-2
Formula:C₈H₁₈O₄
MDL Number:MFCD00008504