Succinyl chloride, Sigma-Aldrich®
Catalog # 8.00678.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 ml
- Seq:2370
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:20 °C
- Molar Mass:154.98 g/mol
- Boiling Point:190 °C
- Flash Point:76 °C
- Refractive Index:1.4691 (20 °C, 589 nm)
- Density:1.38 g/cm3 (20 °C)
Specification Test Results
About this item
Synonyms: Succinyl dichloride
:
:154.98 g/mol
EINECS:208-838-0
UN:3265
:GHS05
Storage Temperature:Ambient
CAS Number:543-20-4
Formula:C₄H₄Cl₂O₂
MDL Number:MFCD00000749
ADR:8 II
:M-Clarity™ quality level = MQ200