N,N-Diisopropylformamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2700-30-3
:
Formula:C₇H₁₅NO
MDL Number:MFCD00008867
:129.2 g/mol
Boiling Pt:195…196 °C
Melting Pt:10…11 °C
Density:0,889
Flash Pt:75 °C (167 °F)
Storage Temperature:Ambient
EINECS:220-284-1
Related Information
Frequently Bought Together