1,2-Epoxydecane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2404-44-6
:
Formula:C₁₀H₂₀O
MDL Number:MFCD00005158
:156.27 g/mol
Boiling Pt:92…93 °C (14 mmHg)
Density:0,840
Flash Pt:78 °C (172 °F)
EINECS:219-295-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together