

N,N'-Dimethyl-1,3-propanediamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-33-1
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00008292
:102.18 g/mol
Boiling Pt:143…146 °C
Density:0,817
Flash Pt:32 °C (90 °F)
Storage Temperature:Ambient
EINECS:203-859-1
ADR:3,II
:GHS02
:GHS05
Related Information
Frequently Bought Together