

Isobutyl methacrylate ≥99.5% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-86-9
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00008931
Boiling Pt:155 °C
Density:0,889
Flash Pt:46 °C (115 °F)
Storage Temperature:Ambient
ADR:3,III
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
:142.2 g/mol
EINECS:202-613-0
UN:2283
:GHS02
:GHS07
Related Information
Frequently Bought Together