

Bis(cyclopentadienyl)titanium dichloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1271-19-8
:
Formula:C₁₀H₁₀Cl₂Ti
MDL Number:MFCD00003723
:248.97 g/mol
Merck Index:14,9482
Melting Pt:∼ 285 °C (dec.)
Density:1,60
Storage Temperature:Ambient
EINECS:215-035-9
ADR:8,II
:Packaged under argon.
UN:3261
:GHS08
:GHS07
Related Information
Frequently Bought Together