

1,4-Butanediol diacrylate ≥99% (reactive esters) stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1070-70-8
:
Formula:C₁₀H₁₄O₄
MDL Number:MFCD00014940
:198.22 g/mol
Boiling Pt:83 °C (0,3 mmHg)
Density:1,051
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:213-979-6
ADR:8,III
:Stabilised with hydroquinone (HQ)
UN:1760
:GHS05
:GHS07
Related Information
Frequently Bought Together