
Phosphinic acid (Hypophosphorous acid) 50% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6303-21-5
:
Formula:H₃O₂P
MDL Number:MFCD02183592
:66 g/mol
Density:1,225
Storage Temperature:Ambient
EINECS:228-601-5
UN:3264
ADR:8,II
:GHS05
Related Information
Frequently Bought Together