Tetraethylene glycol monomethyl ether 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23783-42-8
:
Formula:C₉H₂₀O₅
MDL Number:MFCD00041756
:208.25 g/mol
Boiling Pt:158…160 °C (5 mmHg)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:245-883-5
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4440 to 1.4460 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together