

Isobutyl methacrylate 99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-86-9
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00008931
Melting Pt:–37 °C
Flash Pt:46 °C
Storage Temperature:Ambient
EINECS:202-613-0
UN:2283
ADR:3,III
:142.2 g/mol
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4190 to 1.4220 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥98.5 %
- Stabilizer:15 to 20 ppm MEHQ
- Acidity:≤0.005 % (as methacrylic acid)
Specification Test Results
Frequently Bought Together