(1-Ethoxyethylidene)malononitrile 99+%
:5417-82-3
:
:C₇H₈N₂O
:MFCD00001852
:136.15 g/mol
:89…93 °C
:226-521-5
:3439
:GHS07
:Ambient
- Appearance:White to beige Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:89 to 93 °C
- GC:≥99.0 %
- Sulfated ash:≤1.0 %
- pH:3.5 to 5.5 (c=4, water)
- Specific optical rotation:+107° to +121° (20 °C, 589 nm) (c=1, water)
Frequently Bought Together