Strontium nitrate
Catalog # MOLE42799711-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:211.63 g/mol
Merck Index:13,08926
Boiling Pt:645 °C (1013 hPa)
Melting Pt:570 °C
Density:2,113 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-131-9
UN:1507
ADR:5.1,III
:GHS03
CAS Number:10042-76-9
Formula:Sr(NO₃)₂
MDL Number:MFCD00011248