Order Entry
Northern Ireland
ContactUsLinkComponent
Calcium chloride, Supelco®
Irritant
Calcium chloride, Supelco®
Supplier:  Merck
Certificates
Irritant
Calcium chloride, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    740 g/l (20 °C)
  • Melting Point:
    772 °C
  • Molar Mass:
    110.98 g/mol
  • Boiling Point:
    1600 °C
  • Density:
    2.15 g/cm3 (20 °C)
  • pH:
    8 - 10 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together