

Lithium metavanadate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15060-59-0
:
Formula:LiO₃V
MDL Number:MFCD00053997
:105.88 g/mol
Melting Pt:605 °C
Density:2,74 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:239-124-7
UN:3285
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together