Sulphuric acid 98% ≥98%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
Boiling Pt:330 °C (1013 hPa)
Melting Pt:10.38 °C
Density:1.84 g/cm³ (20 °C)
Molecular Weight:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8 II
REACH:01-2119458838-20
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98.0 %
- Colour:Max. 10 APHA
- Cl (Chloride):Max. 0.00002 %
- NO3 (Nitrate):Max. 0.00002 %
- Al (Aluminium):Max. 0.00001 %
- As (Arsenic):Max. 0.000005 %
- Ba (Barium):Max. 0.000005 %
- Ca (Calcium):Max. 0.00010 %
- Cd (Cadmium):Max. 0.000005 %
- Co (Cobalt):Max. 0.000002 %
- Cr (Chromium):Max. 0.00001 %
- Cu (Copper):Max. 0.000002 %
- Fe (Iron):Max. 0.00010 %
- K (Potassium):Max. 0.00005 %
- Mg (Magnesium):Max. 0.00005 %
- Mn (Manganese):Max. 0.000002 %
- Mo (Molybdenum):Max. 0.000005 %
- NH4 (Ammonium):Max. 0.00020 %
- Na (Sodium):Max. 0.0005 %
- Ni (Nickel):Max. 0.000005 %
- Pb (Lead):Max. 0.000005 %
- Se (Selenium):Max. 0.0001 %
- Sr (Strontium):Max. 0.000002 %
- Zn (Zinc):Max. 0.00005 %
- Substances reducing KMnO4 (as O):Max. 0.0001 %
- Non-volatile matter:Max. 0.002 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC102765G spec_VWRC102766H
Product specification
VWR® Nitrogen Determination According to Kjeldahl
Product Literature
Frequently Bought Together