


Sodium hydrogensulphide, anhydrous
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16721-80-5
:
Formula:HNaS
MDL Number:MFCD00011124
:56.06 g/mol
Merck Index:12,08730
Melting Pt:350 °C
Density:1,79
EINECS:240-778-0
UN:2318
ADR:4.2,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together