

Sodium hexafluorophosphate 98.5+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21324-39-0
:
Formula:F₆NaP
MDL Number:MFCD00011122
Melting Pt:200 °C
Density:2,505 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:244-333-1
UN:3260
ADR:8,II
:167.95 g/mol
:GHS07
:GHS05
Specification Test Results
- Appearance:White to light grey Crystalline powder
- Assay:≥98.50 %
Specification Test Results
Frequently Bought Together