Order Entry
Northern Ireland
ContactUsLinkComponent
Bismuth(III) carbonate basic Blend ZS, Macron Fine Chemicals™
Corrosive
Bismuth(III) carbonate basic Blend ZS, Macron Fine Chemicals™
Catalog # 0288-24
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Bismuth(III) carbonate basic Blend ZS, Macron Fine Chemicals™
Catalog # 0288-24
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    100 lb
  • Seq:
    7167

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance (white to off-white powder):
    Passes Test
  • pH:
    5.0 - 8.5
  • Identification (Zinc Stearate):
    Passes Test
  • Loss on Ignition, For Information Only:
    For Information Only

Specification Test Results

About this item

 :
 :509.97 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:6,86 kg/L
Storage Temperature:Ambient
EINECS:215-185-5
ADR:8,II
REACH:01-2119457892-27
 :GHS05
CAS Number:5892-10-4
Formula:(BiO)₂CO₃
MDL Number:MFCD00010877