Lithium niobate ≥99.998% (metals basis excluding Ta), Puratronic® (Ta <50 ppm)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12031-63-9
:
Formula:LiNbO₃
MDL Number:MFCD00011080
:147.85 g/mol
Melting Pt:∼1275 °C
Density:4,3
Storage Temperature:Ambient
EINECS:234-755-4
Frequently Bought Together