Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium amide for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Sodium amide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
Sodium amide for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    210 °C
  • Molar Mass:
    39.01 g/mol
  • Boiling Point:
    400 °C (1013 hPa)
  • Density:
    1.39 g/cm3 (25 °C)
  • Ignition Point:
    450 °C

Specification Test Results

Related Information

Frequently Bought Together