
Sodium dicyanoamide 97%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1934-75-4
:
Formula:C₂N₃Na
MDL Number:MFCD00003535
Melting Pt:>300 °C (dec.)
:89.03 g/mol
EINECS:217-703-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white to light pink Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
- Water:≤0.5 %
Specification Test Results
Frequently Bought Together