Potassium permanganate 99.0-100.5%, crystallised Ph. Eur.
:7722-64-7
:
:KMnO₄
:MFCD00011364
:158.03 g/mol
:12,07824
:> 240 °C
:2,7 g/cm³ (20 °C)
:231-760-3
:1490
:5.1,II
:GHS03
:GHS07
:GHS09
:Ambient
- Assay:99.0 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification (K):Passes test
- Appearance of solution:Passes test
- Substances insoluble in water:≤ 1.0 %
- Cl (Chloride):≤ 200 ppm
- SO₄ (Sulphate):≤ 500 ppm
- Water:≤ 0.5 %
- Residual solvents:Passes test
- As (Arsenic):≤ 1 ppm
- Cd (Cadmium):≤ 1 ppm
- Co (Cobalt):≤ 1 ppm
- Hg (Mercury):≤ 1 ppm
- Ni (Nickel):≤ 1 ppm
- Pb (Lead):≤ 1 ppm
- V (Vanadium):≤ 1 ppm
- Conforms to Ph.Eur.:Passes test
Frequently Bought Together