ContactUsLinkComponent
Potassium permanganate 99+% ACS
Potassium permanganate 99+% ACS
Certificates
undefined
Potassium permanganate 99+% ACS
Specification Test Results
  • Appearance:
    Dark green-black Glistening crystalline powder
  • Titration with KMnO4:
    ≥99.0 %
  • Chloride and Chlorate (as Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.02 %
  • Insoluble matter:
    ≤0.2 %