


Sodium hexanitritocobaltate(III)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13600-98-1
Formula:Na₃Co(NO₂)₆
MDL Number:MFCD00003512
Merck Index:13,08677
Melting Pt:220 °C (dec.)
Storage Temperature:Ambient
EINECS:237-077-7
UN:1479
ADR:5.1,II
:403.94 g/mol
:GHS03
:GHS07
:GHS08
Specification Test Results
- Appearance:Orange to orange-brown Powder
- Suitability:(for K determination) Passes test
- Insoluble matter:≤0.02 % (in dilute acetic acid)
Specification Test Results
Frequently Bought Together