Watch dishes, BOLA
Dish, made of PTFE, white.
- Convex shape
- Universal chemical resistance
- Temperature resistance: from −200 to +250 °C
For blends or for covering vessels.
Frequently Bought Together
Dish, made of PTFE, white.
For blends or for covering vessels.