


Potassium chlorate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:3811-04-9
:
Formula:KClO₃
MDL Number:MFCD00011361
:122.55 g/mol
Merck Index:13,07703
Boiling Pt:400 °C (1013 hPa)
Melting Pt:356 °C
Density:2,34 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1485
ADR:5.1,II
:M-Clarity™ quality level = MQ300
EINECS:223-289-7
:GHS03
:GHS07
:GHS09
Specification Test Results
- Solubility:73 g/l (20 °C)
- Melting Point:356 °C
- Molar Mass:122.55 g/mol
- Bulk Density:1200 - 1400 kg/m3
- Boiling Point:400 °C (decomposition)
- Vapor Pressure:(20 °C)
- Density:2.32 g/cm3 (20 °C)
- pH:5.6 (73 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Related Information
Frequently Bought Together