Sodium carbonate, anhydrous 99.5-100.5% Ph. Eur., NF, BP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
:105.99 g/mol
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-838-8
REACH:01-2119485498-19
:GHS07
Specification Test Results
- Appearance:Conforms (see CoA/CoS)
- Heavy metals (as Pb):≤ 50 ppm
- Residual solvents:Unlikely by manuf.process
- Residues of metal catalysts or reagents:Unlikely by manuf.process
- As (Arsenic):≤ 5 ppm
- Cd (Cadmium):≤ 1 ppm
- Co (Cobalt):≤ 1 ppm
- Hg (Mercury):≤ 2 ppm
- Ni (Nickel):≤ 1 ppm
- Pb (Lead):≤ 5 ppm
- V (Vanadium):≤ 6 ppm
- Conforms to Ph.Eur./NF/BP:Passes test
- Ph.Eur./BP specifications::.
- Assay (dried substance) [Ph.Eur./BP]:99.5 - 100.5 %
- Identification A:Passes test Ph.Eur./BP
- Identification B:Passes test Ph.Eur./BP
- Identification C:Passes test Ph.Eur./BP
- Identification D [Ph.Eur./BP]:≤ 1.0 %
- Appearance of solution:Passes test Ph.Eur./BP
- Alkali hydroxides and bicarbonates:Passes test Ph.Eur./BP
- Cl (Chloride) [Ph.Eur./BP]:≤ 125 ppm
- SO4 (Sulphate) [Ph.Eur./BP]:≤ 250 ppm
- Fe (Iron) [Ph.Eur./BP]:≤ 50 ppm
- Loss on drying (300 ± 15°C):≤ 1.0 %
- NF specifications::.
- Assay (calculated on anhydrous) [NF]:99.5 - 100.5 %
- Water determination [NF]:≤ 0.5 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 50L Plastic bucket UN Y (ZPACK13247050)
Packaging Information
27767_SODIUM CARBONATE ANHYDROUS PH.EUR._PHTALATE_REV0.pdf Sodium Carbonate, Anhydrous 99,5 - 100,5% Ph. Eur., NF, BP
Additional Product Details
spec_VWRC27767.295 spec_VWRC27767.466
Product specification
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together