
Lithium metaborate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13453-69-5
:
Formula:BLiO₂
MDL Number:MFCD00011089
:49.75 g/mol
Merck Index:13,05546
Boiling Pt:1719 °C (1013 hPa)
Melting Pt:845 °C
Density:1,397
EINECS:236-631-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together