


Lithium tetrahydroborate ≥95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16949-15-8
:
Formula:H₄BLi
MDL Number:MFCD00011088
:21.78 g/mol
Merck Index:14,5525
Boiling Pt:66 °C (1013 hPa)
Melting Pt:∼ 280 °C (dec.)
Density:0,66
Storage Temperature:Ambient
EINECS:241-021-7
UN:1413
ADR:4.3,I
:GHS02
:GHS06
:GHS05
Frequently Bought Together