
1,5-Diazobicyclo[4.3.0]-5-nonene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3001-72-7
:
Formula:C₇H₁₂N₂
MDL Number:MFCD00005554
:124.19 g/mol
Boiling Pt:95…98 °C (7,5 mmHg)
Density:1 g/cm³
Flash Pt:94 °C
Storage Temperature:Ambient
EINECS:221-087-3
UN:3267
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5190 to 1.5210 (20°C, 589 nm)
- Water:≤1 %
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together