Order Entry
Northern Ireland
ContactUsLinkComponent
Methylcyclohexane for synthesis, Sigma-Aldrich®
Flammable
Health
Irritant
Environment
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Health
Irritant
Environment
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.014 g/l (20 °C)
  • Melting Point:
    -126 °C
  • Molar Mass:
    98.19 g/mol
  • Boiling Point:
    100.9 °C (1013 hPa)
  • Vapor Pressure:
    48 hPa (20 °C)
  • Flash Point:
    -4 °C
  • Explosion Limit:
    1.1 - 6.7 %(V)
  • Density:
    0.77 g/cm3 (20 °C)
  • Ignition Point:
    260 °C
  • Hill Formula:
    C7H14
  • Chemical Formula:
    C6H11CH3

Specification Test Results

Related Information

Frequently Bought Together