2-Acetylcyclohexanone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:874-23-7
:
Formula:C₈H₁₂O₂
MDL Number:MFCD00001633
:140.18 g/mol
Density:1,063 g/cm³
Flash Pt:79 °C
Storage Temperature:Ambient
EINECS:212-858-5
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5070 to 1.5130 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together