Organic reference standard, Bis(pentafluorophenyl)phenylphosphine 1,000 µg/ml in acetone, Supelco®
Catalog # 47941.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- CAS No.:5074-71-5
- Description:Bis(pentafluorophenyl)phenylphosphine 1,000 µg/ml in acetone
- Formula:C₁₈H₅F₁₀P
- Description:Bis(pentafluorophenyl)phenylphosphine