Order Entry
Northern Ireland
ContactUsLinkComponent
Bis(pentafluorophenyl)phenylphosphine
Bis(pentafluorophenyl)phenylphosphine
Catalog # 442543.
Supplier:  Merck
CAS Number:  
undefined
Bis(pentafluorophenyl)phenylphosphine
Catalog # 442543.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    2440
  • Pk Info:
    100 mg

Specifications

About this item

 :
 :442.2 g/mol
EINECS:225-780-1
CAS Number:5074-71-5
Formula:C₁₈H₅F₁₀P
MDL Number:MFCD00000291