Saccharin
Catalog # PROOICRS0411
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:183.19 g/mol
EINECS:201-321-0
CAS Number:81-07-2
Formula:C₇H₅NO₃S
MDL Number:MFCD00005866