Saccharin, Sigma-Aldrich®
: Merck
- Solubility:(20 °C) insoluble
- Melting Point:228 - 230 °C
- Molar Mass:183.18 g/mol
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Contact slides
: VWR Chemicals
Benzanilide ≥98%
: Thermo Fisher Scientific
1-Acetyl-2-phenylhydrazine ≥98%
: Thermo Fisher Scientific