Order Entry
Northern Ireland
ContactUsLinkComponent
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Millipore®
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Millipore®
Catalog # 394675-1
CAS Number:  
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Millipore®
Catalog # 394675-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :243.28 g/mol
Storage Temperature:Ambient
EINECS:249-954-1
CAS Number:29915-38-6
Formula:C₇H₁₇NO₆S
MDL Number:MFCD00007538